Learn More
Medchemexpress LLC Cediranib maleate | 857036-77-2 | 99.9% | 50 MG
SDP
Catalog No. 5000340325
Encompass_Preferred
Cediranib maleate is a highly potent, orally available VEGFR inhibitor. It effectively inhibits VEGF-stimulated proliferation and KDR phosphorylation in human umbilical vein endothelial cells and reduces vessel sprouting in coculture models. In vivo, it ablates experimental angiogenesis and inhibits the growth of various human tumor xenografts.
  • Highly potent, orally available VEGFR inhibitor
  • Inhibits VEGF-stimulated proliferation and KDR phosphorylation
  • Reduces vessel area, length, and branching at subnanomolar concentrations
  • Ablates experimental angiogenesis
  • Inhibits growth of established human tumor xenografts
Catalog No. 50-003-40325 Supplier Medchemexpress LLC Supplier No. HY1304950MG
Cediranib maleate is a highly potent, orally available VEGFR inhibitor. It effectively inhibits VEGF-stimulated proliferation and KDR phosphorylation in human umbilical vein endothelial cells and reduces vessel sprouting in coculture models. In vivo, it ablates experimental angiogenesis and inhibits the growth of various human tumor xenografts.
  • Highly potent, orally available VEGFR inhibitor
  • Inhibits VEGF-stimulated proliferation and KDR phosphorylation
  • Reduces vessel area, length, and branching at subnanomolar concentrations
  • Ablates experimental angiogenesis
  • Inhibits growth of established human tumor xenografts