Learn More
Medchemexpress LLC Cediranib maleate | 857036-77-2 | 99.95% | 100 MG
SDP
Catalog No. 5000338616
Encompass_Preferred
Cediranib maleate is a highly potent, orally available VEGFR inhibitor. It inhibits VEGF-stimulated proliferation and KDR phosphorylation, reducing vessel area, length, and branching. It ablates experimental angiogenesis and inhibits endochondral ossification and corpora luteal development. It also inhibits the growth of established human tumor xenografts in athymic mice, showing a reduction in microvessel density within tumors.
  • Highly potent, orally available VEGFR inhibitor
  • Inhibits VEGF-stimulated proliferation and KDR phosphorylation
  • Reduces vessel area, length, and branching
  • Ablates experimental angiogenesis
  • Inhibits endochondral ossification and corpora luteal development
  • Inhibits growth of established human tumor xenografts
  • Reduces microvessel density within tumors
Catalog No. 50-003-38616 Supplier Medchemexpress LLC Supplier No. HY13049100MG
Cediranib maleate is a highly potent, orally available VEGFR inhibitor. It inhibits VEGF-stimulated proliferation and KDR phosphorylation, reducing vessel area, length, and branching. It ablates experimental angiogenesis and inhibits endochondral ossification and corpora luteal development. It also inhibits the growth of established human tumor xenografts in athymic mice, showing a reduction in microvessel density within tumors.
  • Highly potent, orally available VEGFR inhibitor
  • Inhibits VEGF-stimulated proliferation and KDR phosphorylation
  • Reduces vessel area, length, and branching
  • Ablates experimental angiogenesis
  • Inhibits endochondral ossification and corpora luteal development
  • Inhibits growth of established human tumor xenografts
  • Reduces microvessel density within tumors