Learn More
Medchemexpress LLC Cediranib maleate | 857036-77-2 | 99.95% | 25 MG
SDP
Catalog No. 5000341768
Encompass_Preferred
Cediranib maleate is a highly potent, orally available VEGFR inhibitor. It demonstrates inhibitory activity against Flt1, KDR, Flt4, PDGFRα, PDGFRβ, and c-Kit. This compound is intended for research use only.
  • Inhibits VEGF-stimulated proliferation and KDR phosphorylation in human umbilical vein endothelial cells.
  • Reduces vessel area, length, and branching in a fibroblast/endothelial cell coculture model.
  • Ablates experimental (VEGF-induced) angiogenesis in vivo.
  • Inhibits endochondral ossification and corpora luteal development.
  • Dose-dependently inhibits the growth of human tumor xenografts (colon, lung, prostate, breast, and ovary) in athymic mice.
  • Shows a reduction in microvessel density within tumors, indicating vascular regression.
Catalog No. 50-003-41768 Supplier Medchemexpress LLC Supplier No. 25MGHY13049
Cediranib maleate is a highly potent, orally available VEGFR inhibitor. It demonstrates inhibitory activity against Flt1, KDR, Flt4, PDGFRα, PDGFRβ, and c-Kit. This compound is intended for research use only.
  • Inhibits VEGF-stimulated proliferation and KDR phosphorylation in human umbilical vein endothelial cells.
  • Reduces vessel area, length, and branching in a fibroblast/endothelial cell coculture model.
  • Ablates experimental (VEGF-induced) angiogenesis in vivo.
  • Inhibits endochondral ossification and corpora luteal development.
  • Dose-dependently inhibits the growth of human tumor xenografts (colon, lung, prostate, breast, and ovary) in athymic mice.
  • Shows a reduction in microvessel density within tumors, indicating vascular regression.