Learn More
Medchemexpress LLC Cediranib maleate | 857036-77-2 | MFCD09954115 | 100.0% | C29H31FN4O7 | 10MG
SDP
Catalog No. 5000260172
Encompass_Preferred
Cediranib maleate is the maleate salt of cediranib (AZD-2171), a potent orally available vascular endothelial growth factor receptor (VEGFR) tyrosine kinase inhibitor supplied for research use. It is provided as a high-purity solid and is also available in ready-to-use DMSO stock solutions for biochemical and cell-based assays.
  • High purity 99.95%.
  • Potent VEGFR inhibitor with low-nanomolar activity against Flt1, KDR, Flt4, PDGFRα/β, and c-Kit.
  • Suitable for in vitro and in vivo research applications.
  • Supplied as a solid and as 10 mM DMSO solutions.
  • Molecular formula C29H31FN4O7; molecular weight 566.58 Da.
Catalog No. 50-002-60172 Supplier Medchemexpress LLC Supplier No. HY1304910MG
Cediranib maleate is the maleate salt of cediranib (AZD-2171), a potent orally available vascular endothelial growth factor receptor (VEGFR) tyrosine kinase inhibitor supplied for research use. It is provided as a high-purity solid and is also available in ready-to-use DMSO stock solutions for biochemical and cell-based assays.
  • High purity 99.95%.
  • Potent VEGFR inhibitor with low-nanomolar activity against Flt1, KDR, Flt4, PDGFRα/β, and c-Kit.
  • Suitable for in vitro and in vivo research applications.
  • Supplied as a solid and as 10 mM DMSO solutions.
  • Molecular formula C29H31FN4O7; molecular weight 566.58 Da.