Learn More
Selleck Chemical LLC Cedryl acetate S4793-25mg
SDP
Catalog No. 503112022
Encompass_Preferred
Cedryl Acetate (Cedrol acetate Cedranyl acetate) an acetylated from cedarwood oil has been applied to chemistry for its properties as a chiral and cell signaling reagent with antifungal and immunotoxicity functions Cedryl acetate exhibits -glucosidase inhibitory activity
Catalog No. 50-311-2022 Supplier Selleck Chemical LLC Supplier No. S479325MG
Cedryl Acetate (Cedrol acetate Cedranyl acetate) an acetylated from cedarwood oil has been applied to chemistry for its properties as a chiral and cell signaling reagent with antifungal and immunotoxicity functions Cedryl acetate exhibits -glucosidase inhibitory activity