Learn More
Medchemexpress LLC Cedryl acetate | 77-54-3 | MFCD00077764 | 99.8% | 264.40 | 100 MG
SDP
Catalog No. 5000453738
Encompass_Preferred
Cedryl acetate is a tricyclic sesquiterpene isolated from the plant *Cunninghamia lanceolata*. It shows α-glucosidase inhibitory activity.
  • Isolated from the plant *Cunninghamia lanceolata*.
  • Shows α-glucosidase inhibitory activity.
  • For research use only.
Catalog No. 50-004-53738 Supplier Medchemexpress LLC Supplier No. HYW009417100MG
Cedryl acetate is a tricyclic sesquiterpene isolated from the plant *Cunninghamia lanceolata*. It shows α-glucosidase inhibitory activity.
  • Isolated from the plant *Cunninghamia lanceolata*.
  • Shows α-glucosidase inhibitory activity.
  • For research use only.