Learn More
Medchemexpress LLC HY-101453 5mg Medchemexpress, Ceefourin 1 CAS:315702-40-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1014535MG
Encompass_Preferred
Medchemexpress, HY-101453 5mg Ceefourin 1 CAS:315702-40-0 Ceefourin 1 is a potent and highly selective multidrug resistance protein 4 (MRP4) inhibitor. Ceefourin 1 inhibits transport of a broad range of MRP4 substrates, yet is highly selective for MRP4 over other ABC transporters. Ceefourin 1 is a benzothiazol and primarily as a chemosensitizer for high-risk neuroblastoma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0634