Learn More
STEMCELL Technologies CEF (HLA Class I Control) Peptide Pool, Size: 1 Unit
SDP
Catalog No. NC2133298
Encompass
The CEF (HLA Class I Control) Peptide Pool is a lyophilized mixture of 32 peptides from cytomegalovirus (CMV), Epstein-Barr virus (EBV), and influenza virus. The pool consists of defined HLA class I-restricted T cell epitopes from these three viruses, and can be used as a positive control to stimulate T cells.
This item is not returnable. View return policy
Catalog No. NC2133298 Supplier STEMCELL Technologies Supplier No. 1000675
Add to Cart
Add to Cart
The CEF (HLA Class I Control) Peptide Pool is a lyophilized mixture of 32 peptides from cytomegalovirus (CMV), Epstein-Barr virus (EBV), and influenza virus. The pool consists of defined HLA class I-restricted T cell epitopes from these three viruses, and can be used as a positive control to stimulate T cells.