Learn More
Medchemexpress LLC Cef20 TFA | 99.3% | 1057.19 | 5 MG
SDP
Catalog No. 5000423200
Encompass_Preferred
CEF20 TFA is an HLA-A*0201-restricted epitope from cytomegalovirus pp65 (495-503). It is for research use only.
  • An HLA-A*0201-restricted epitope from cytomegalovirus pp65 (495-503)
  • For research use only
Catalog No. 50-004-23200 Supplier Medchemexpress LLC Supplier No. HYP1780A5MG
CEF20 TFA is an HLA-A*0201-restricted epitope from cytomegalovirus pp65 (495-503). It is for research use only.
  • An HLA-A*0201-restricted epitope from cytomegalovirus pp65 (495-503)
  • For research use only