Learn More
Medchemexpress LLC Cefaclor | 53994-73-3 | 98.5% | 1 ML
SDP
Catalog No. 5000407304
Encompass_Preferred
Cefaclor is a well-absorbed orally active cephalosporin antibiotic that specifically binds to penicillin-binding protein 3 (PBP3). It is used in research for various bacterial infections, including respiratory tract infections, bacterial bronchitis, pharyngitis, and skin infections, and also for depression studies.
  • Specifically binds to penicillin-binding protein 3 (PBP3)
  • Used for research on depression
  • Active against various bacterial infections, such as respiratory tract infections, bacterial bronchitis, pharyngitis, and skin infections
  • Well-absorbed orally active cephalosporin antibiotic
Catalog No. 50-004-07304 Supplier Medchemexpress LLC Supplier No. HYB019810MM/1ML
Cefaclor is a well-absorbed orally active cephalosporin antibiotic that specifically binds to penicillin-binding protein 3 (PBP3). It is used in research for various bacterial infections, including respiratory tract infections, bacterial bronchitis, pharyngitis, and skin infections, and also for depression studies.
  • Specifically binds to penicillin-binding protein 3 (PBP3)
  • Used for research on depression
  • Active against various bacterial infections, such as respiratory tract infections, bacterial bronchitis, pharyngitis, and skin infections
  • Well-absorbed orally active cephalosporin antibiotic