Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Cefaclor, 96%

Catalog No. AC455260010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455260010 1 g
AC455260050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC455260010 Supplier Thermo Scientific Chemicals Supplier No. 455260010
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53994-73-3
Molecular Formula C15H14ClN3O4S
Molecular Weight (g/mol) 367.804
InChI Key QYIYFLOTGYLRGG-GPCCPHFNSA-N
PubChem CID 51039
ChEBI CHEBI:3478
IUPAC Name (6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES C1C(=C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)Cl

Specifications

Type Cefaclor
Infrared Spectrum Authentic
Water 3.0 to 9.0%
Quantity 1 g
Formula Weight 367.81
Percent Purity 95.0% (HPLC)
Chemical Name or Material Cefaclor
Hazard Category Danger
Hazard Statement GHS H Statement May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing. IF ON SKIN: Wash with plenty of soap and water. If skin irritation or rash occurs: Get medical advice/attention. In case of inadequate ventilation wear respiratory protection. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.