Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefaclor, ∽98%, MP Biomedicals™

Catalog No. ICN19894380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19894380 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19894380 Supplier MP Biomedicals, Inc Supplier No. 0219894380

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 53994-73-3
Molecular Formula C15H14ClN3O4S
Molecular Weight (g/mol) 367.804
MDL Number MFCD00151471
InChI Key QYIYFLOTGYLRGG-GPCCPHFNSA-N
Synonym cefaclor, ceclor, cephaclor, cefaclorum, cefaclor anhydrous, kefral, raniclor, panoral, alfacet, distaclor
PubChem CID 51039
ChEBI CHEBI:3478
IUPAC Name (6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES C1C(=C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)Cl

Specifications

Color Yellow
Assay Percent Range ∽98%
Quantity 100 mg
Formula Weight 367.8
Percent Purity ∼98%
Physical Form Powder
Chemical Name or Material Cefaclor
Recommended Storage Store at Room Temperature (15 to 30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.