Learn More
MOLECULAR DEPOT LLC Cefaclor BSA - 1 mg
Catalog No. 5000013440
Encompass_Preferred
Cefaclor BSA conjugate is formed between cefaclor an antibiotic and bovine serum albumin (BSA) This conjugate enhances the immune response to cefaclor allowing for the generation of antibodies that can be used in various research applications For in vitro research use only
Catalog No. 50-000-13440 Supplier MOLECULAR DEPOT LLC Supplier No. B2020844
Cefaclor BSA conjugate is formed between cefaclor an antibiotic and bovine serum albumin (BSA) This conjugate enhances the immune response to cefaclor allowing for the generation of antibodies that can be used in various research applications For in vitro research use only