Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefaclor, G-Biosciences
SCI_ED

Catalog No. S139748
Change view
Click to view available options
Quantity:
500 mg
1000 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139748 1000 mg
S139089 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139748 Supplier G-Biosciences Supplier No. RC909
Add to Cart
Add to Cart

Cefaclor is a second generation cephalosporin antibiotic isolated from the Acremonium fungus. It targets a wide variety of bacteria including Streptococcus pneumoniae and Haemophilus influenzae and a variety of other gram-negative and gram-positive bacteria.

Cefaclor has been useful in studying urinary tract and intra-abdominal infections.

Chemical Identifiers

CAS 53994-73-3
Molecular Weight (g/mol) 367.81 g/mol

Specifications

Technique HPLC, Spectrometry, UV/Vis, Ion Chromatography, Mass Spectrometry
Certifications/Compliance ISO, ISO 9001, REACH, GHS/OSHA, NIST
Type Drug Standard
Element Multi-element
Industry Type Biotech
Quantity 1000 mg
Percent Purity >97%
Grade Molecular Biology
Chemical Name or Material Cefaclor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.