Learn More
Medchemexpress LLC Cefaclor monohydrate | 70356-03-5 | 99.1% | 100 MG
SDP
Catalog No. 5000405791
Encompass_Preferred
Cefaclor is an orally active cephalosporin antibiotic that is well-absorbed. It specifically binds to penicillin-binding protein 3 (PBP3). This antibiotic is utilized in research for studying depression and various bacterial infections, such as respiratory tract infections, bacterial bronchitis, pharyngitis, and skin infections.
  • Orally active cephalosporin antibiotic
  • Well-absorbed
  • Binds to penicillin-binding protein 3 (PBP3)
  • Used in research for depression and bacterial infections
  • Exhibits antibacterial activity against Gram-positive and Gram-negative isolates
  • Transported by hPepT2 and hPepT1
Catalog No. 50-004-05791 Supplier Medchemexpress LLC Supplier No. HYB0198A100MG
Cefaclor is an orally active cephalosporin antibiotic that is well-absorbed. It specifically binds to penicillin-binding protein 3 (PBP3). This antibiotic is utilized in research for studying depression and various bacterial infections, such as respiratory tract infections, bacterial bronchitis, pharyngitis, and skin infections.
  • Orally active cephalosporin antibiotic
  • Well-absorbed
  • Binds to penicillin-binding protein 3 (PBP3)
  • Used in research for depression and bacterial infections
  • Exhibits antibacterial activity against Gram-positive and Gram-negative isolates
  • Transported by hPepT2 and hPepT1