Learn More
Medchemexpress LLC Cefaclor monohydrate | 70356-03-5 | 99.1% | 500 MG
SDP
Catalog No. 5000407386
Encompass_Preferred
Cefaclor is a well-absorbed, orally active cephalosporin antibiotic. It specifically binds to penicillin-binding protein 3 (PBP3) and can be used in research for depression and various bacterial infections, including respiratory tract infections, bacterial bronchitis, pharyngitis, and skin infections.
  • Well-absorbed, orally active cephalosporin antibiotic.
  • Specifically binds to penicillin-binding protein 3 (PBP3).
  • Used for research of depression and various bacterial infections.
  • Effective against respiratory tract infections, bacterial bronchitis, pharyngitis, and skin infections.
  • Exhibits antibacterial activity against Gram-positive and Gram-negative isolates.
  • Transported by hPepT2 and hPepT1 in MDCK cells.
Catalog No. 50-004-07386 Supplier Medchemexpress LLC Supplier No. HYB0198A500MG
Cefaclor is a well-absorbed, orally active cephalosporin antibiotic. It specifically binds to penicillin-binding protein 3 (PBP3) and can be used in research for depression and various bacterial infections, including respiratory tract infections, bacterial bronchitis, pharyngitis, and skin infections.
  • Well-absorbed, orally active cephalosporin antibiotic.
  • Specifically binds to penicillin-binding protein 3 (PBP3).
  • Used for research of depression and various bacterial infections.
  • Effective against respiratory tract infections, bacterial bronchitis, pharyngitis, and skin infections.
  • Exhibits antibacterial activity against Gram-positive and Gram-negative isolates.
  • Transported by hPepT2 and hPepT1 in MDCK cells.