Learn More
Medchemexpress LLC Cefadroxil | 50370-12-2 | 99.54% | 250 MG
SDP
Catalog No. 5000414326
Encompass_Preferred
Cefadroxil is an orally active broad-spectrum cephalosporin antibiotic. It inhibits bacterial cell wall synthesis and enhances the expression of glutamate transporter-1. Its small intestinal absorption is dependent on the intestinal peptide transporter PepT1. Cefadroxil exhibits inhibitory and bactericidal activity against various Gram-positive and Gram-negative bacteria and also possesses analgesic effects on neuropathic pain.
  • Inhibits bacterial cell wall synthesis
  • Enhances glutamate transporter-1 expression
  • Active against Gram-positive and Gram-negative bacteria
  • Exhibits analgesic effects on neuropathic pain
Catalog No. 50-004-14326 Supplier Medchemexpress LLC Supplier No. 250MGHYB1190
Cefadroxil is an orally active broad-spectrum cephalosporin antibiotic. It inhibits bacterial cell wall synthesis and enhances the expression of glutamate transporter-1. Its small intestinal absorption is dependent on the intestinal peptide transporter PepT1. Cefadroxil exhibits inhibitory and bactericidal activity against various Gram-positive and Gram-negative bacteria and also possesses analgesic effects on neuropathic pain.
  • Inhibits bacterial cell wall synthesis
  • Enhances glutamate transporter-1 expression
  • Active against Gram-positive and Gram-negative bacteria
  • Exhibits analgesic effects on neuropathic pain