Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefadroxil, G-Biosciences
SCI_ED

Catalog No. S157224
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S157224 5 g
S139749 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S157224 Supplier G-Biosciences Supplier No. RC911
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cefadroxil is a first generation cephalosporin antibiotic with a structure similar to that of cephalexin and cephradine.

Cefadroxil is a first generation cephalosporin antibiotic with a structure similar to that of cephalexin and cephradine. It targets a wide variety of bacterial species including Staphylococcus and Streptococcus species.

Chemical Identifiers

Molecular Weight (g/mol) 363.39

Specifications

Physical Form Powder
Quantity 5 g
Product Line Gibco
Boiling Point 789.9°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.