Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefadroxil, MilliporeSigma™ Supelco™

Catalog No. 111014362 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4362 1 g
11-101-4363 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-4362 Supplier MilliporeSigma Supelco Supplier No. C70201G
Add to Cart
Add to Cart

Cefadroxil is a broad spectrum cephalosporin antibiotic, widely used in veterinery medicine for the oral treatment of infectious diseases.

Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 50370-12-2
Physical Form Powder/Crystals
Linear Formula C16H17N3O5 S
MDL Number MFCD00865091
Type Analytical Standard
Quantity 1 g
Formula Weight 363.39
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.