Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefadroxil, MP Biomedicals™

Catalog No. ICN15058105 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15058105 5 g
ICN15058101 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15058105 Supplier MP Biomedicals, Inc Supplier No. 0215058105
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50370-12-2
Molecular Formula C16H17N3O5S
Molecular Weight (g/mol) 363.388
MDL Number MFCD00865091
InChI Key BOEGTKLJZSQCCD-UEKVPHQBSA-N
Synonym p-hydroxycephalexine, Cefamox
PubChem CID 47965
ChEBI CHEBI:3479
IUPAC Name (6R,7R)-7-[[(2R)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CC1=C(N2C(C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)SC1)C(=O)O

Specifications

Type Cefadroxil
Physical Form Crystalline Powder
Color White
Quantity 5 g
Formula Weight 363.4
Grade Not Available
Chemical Name or Material Cefadroxil
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.