Learn More
Cayman Chemical Cefadroxil (hydrate), Purity: ≥98%, CAS: 66592-87-8, 500 mg
SDP
Catalog No. 502984626
Encompass_Preferred
A semisynthetic cephalosporin antibiotic; active against B. anthracis, V. cholerae, and P. multocida strains (MICs = 1, 2, and 4 µg/ml, respectively), among others; protective against infection by strains of S. pyogenes, S. pneumoniae, or S. aureus in mice (PD50s = 0.6-2.4, 18-24, and 0.3-29 mg/kg, respectively)
Catalog No. 50-298-4626 Supplier Cayman Chemical Supplier No. 20365500 MG
A semisynthetic cephalosporin antibiotic; active against B. anthracis, V. cholerae, and P. multocida strains (MICs = 1, 2, and 4 µg/ml, respectively), among others; protective against infection by strains of S. pyogenes, S. pneumoniae, or S. aureus in mice (PD50s = 0.6-2.4, 18-24, and 0.3-29 mg/kg, respectively)