Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefamandole Nafate, G-Biosciences
SCI_ED

Catalog No. S139841
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139841 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139841 Supplier G-Biosciences Supplier No. RC912
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cefamandole nafate is a second generation cephalosporin antibiotic.

It is effective against a variety of gram-negative and gram-positive bacteria. It has been used to study the expression and inhibition of the penicillin binding proteins, especially PBP 2A.

Chemical Identifiers

Molecular Weight (g/mol) 512.49

Specifications

Physical Form Powder
Quantity 5 g
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.