Learn More
Cayman Chemical Cefamandole (sodium salt), Purity: ≥90%, CAS: 30034-03-8, 100 mg
SDP
Catalog No. 502966685
Encompass_Preferred
A cephalosporin antibiotic that is effective against E. coli (MIC values range from 0.25-2 mg/L depending on strain) as well as H. influenza and S. aureus
Catalog No. 50-296-6685 Supplier Cayman Chemical Supplier No. 18138100 MG
A cephalosporin antibiotic that is effective against E. coli (MIC values range from 0.25-2 mg/L depending on strain) as well as H. influenza and S. aureus