Learn More
Medchemexpress LLC HY-107329 10mg Medchemexpress, Cefathiamidine CAS:33075-00-2 Purity:>98%
SDP
Catalog No. 501968380
Encompass_Preferred
Medchemexpress, HY-107329 10mg Cefathiamidine CAS:33075-00-2 Cefathiamidine is a first-generation cephalosporin antibacterial agent and is used to treat infections caused by susceptible bacteria. Cefathiamidine exhibits a wide spectrum of antimicrobial activity against bacteria. Cefathiamidine is used for the treatment of respiratory, liver, five senses, urinary tract infections, endocarditis and sepsis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8380 Supplier Medchemexpress LLC Supplier No. HY10732910MG
Medchemexpress, HY-107329 10mg Cefathiamidine CAS:33075-00-2 Cefathiamidine is a first-generation cephalosporin antibacterial agent and is used to treat infections caused by susceptible bacteria. Cefathiamidine exhibits a wide spectrum of antimicrobial activity against bacteria. Cefathiamidine is used for the treatment of respiratory, liver, five senses, urinary tract infections, endocarditis and sepsis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.