Learn More
Cayman Chemical Cefazolin-13C2,15N, Purity: ≥95%, CAS: 2101505-58-0, 1 mg
SDP
Catalog No. 502932550
Encompass_Preferred
An internal standard for the quantification of cefazolin by GC- or LC-MS
Catalog No. 50-293-2550 Supplier Cayman Chemical Supplier No. 253191 MG
An internal standard for the quantification of cefazolin by GC- or LC-MS