Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cefazolin | 25953-19-9 | 99.7% | 100 MG
SDP

Catalog No. 5000409695
Encompass_Preferred

Cefazolin (Cephazolin) is a first-generation cephalosporin antibiotic that can be used in research for various bacterial infections. It also exhibits anti-inflammatory effects and can mitigate post-operative cognitive dysfunction (POCD).

  • Antibiotic properties: First-generation cephalosporin antibiotic effective against various bacterial infections.
  • Anti-inflammatory effect: Can attenuate post-operative cognitive dysfunction (POCD).
  • High purity of 99.73%.
  • Research use only.
  • Versatile forms: Available as a solid and in solution.

Catalog No. 50-004-09695 Supplier Medchemexpress LLC Supplier No. HYB1892100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cefazolin (Cephazolin) is a first-generation cephalosporin antibiotic that can be used in research for various bacterial infections. It also exhibits anti-inflammatory effects and can mitigate post-operative cognitive dysfunction (POCD).

  • Antibiotic properties: First-generation cephalosporin antibiotic effective against various bacterial infections.
  • Anti-inflammatory effect: Can attenuate post-operative cognitive dysfunction (POCD).
  • High purity of 99.73%.
  • Research use only.
  • Versatile forms: Available as a solid and in solution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.