Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefazolin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013763 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3763 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3763 Supplier MilliporeSigma Supelco Supplier No. PHR1291500MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cefazolin is a first-generation cephalosporin β-lactam antibiotic drug that exhibits broad-spectrum antimicrobial activity against Klebsiella pneumoniae. It also acts in vitro against Gram-positive aerobic cocci while shows only a limited action on Gram-negative bacteria.Pharmaceutical secondary standard for application in quality control. Provides pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
TRUSTED_SUSTAINABILITY

Specifications

CAS 25953-19-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C14H14N8O4S3
Synonym Cefazolin; Cephazolin
Type Certified Reference Material
Quantity 500 mg
Formula Weight 454.51
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.