Learn More
Selleck Chemical LLC Cefazolin Sodium S4595-100mg
SDP
Catalog No. 503108676
Encompass_Preferred
Cefazolin Sodium (cefazoline cephazolin Ancef) is a semisynthetic cephalosporin analog with broad-spectrum antibiotic action due to inhibition of bacterial cell wall synthesis It attains high serum levels and is excreted quickly via the urine
Catalog No. 50-310-8676 Supplier Selleck Chemical LLC Supplier No. S4595100MG
Add to Cart
Add to Cart
Cefazolin Sodium (cefazoline cephazolin Ancef) is a semisynthetic cephalosporin analog with broad-spectrum antibiotic action due to inhibition of bacterial cell wall synthesis It attains high serum levels and is excreted quickly via the urine