Learn More
Apexbio Technology LLC Cefazolin (sodium salt) 27164-46-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000091450
Encompass_Preferred
Cefazolin sodium salt (CAS 27164-46-1) is a semisynthetic antibiotic that inhibits bacterial cell wall synthesis by binding to specific penicillin-binding proteins thereby blocking transpeptidation reactions essential for peptidoglycan formation In in vitro studies cefazolin sodium salt suppresses the proliferation of cultured osteoblast-like MG-63 cells exhibiting substantial cytotoxicity at concentrations of 10 000 g/ml precise IC50 values depend on cell type and experimental conditions Cefazolin sodium salt is commonly used in microbiology pharmacokinetic investigations and infection models particularly within studies of antimicrobial susceptibility and renal dysfunction
Catalog No. 50-000-91450 Supplier Apexbio Technology LLC Supplier No. C41835.1
Cefazolin sodium salt (CAS 27164-46-1) is a semisynthetic antibiotic that inhibits bacterial cell wall synthesis by binding to specific penicillin-binding proteins thereby blocking transpeptidation reactions essential for peptidoglycan formation In in vitro studies cefazolin sodium salt suppresses the proliferation of cultured osteoblast-like MG-63 cells exhibiting substantial cytotoxicity at concentrations of 10 000 g/ml precise IC50 values depend on cell type and experimental conditions Cefazolin sodium salt is commonly used in microbiology pharmacokinetic investigations and infection models particularly within studies of antimicrobial susceptibility and renal dysfunction