Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefazolin Sodium Salt, MP Biomedicals™

Catalog No. ICN15494680 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15494680 100 mg
ICN15494690 500 mg
ICN15494691 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15494680 Supplier MP Biomedicals, Inc Supplier No. 0215494680
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 27164-46-1 | Formula: C14H16N8NaO4S3 | Weight: 479.504 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27164-46-1
Molecular Formula C14H16N8NaO4S3
Molecular Weight (g/mol) 479.504
InChI Key MTIAAUXSENDLGW-SLNAEPSVSA-N
Synonym Acef, Cefazil
PubChem CID 131673922
IUPAC Name (6R,7R)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium
SMILES [HH].CC1=NN=C(S1)SCC2=C(N3C(C(C3=O)NC(=O)CN4C=NN=N4)SC2)C(=O)O.[Na]

Specifications

Type Cefazolin Sodium Salt
Physical Form Powder
Color White
Quantity 100 mg
Formula Weight 476.48
Grade Not Available
Chemical Name or Material Cefazolin Sodium Salt
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.