Learn More
Cayman Chemical Cefazolin (sodium salt), Purity: ≥98%, CAS: 27164-46-1, 500 mg
SDP
Catalog No. 502968921
Encompass_Preferred
A cephalosporin antibiotic; active in vitro against various Gram-positive and Gram-negative bacteria (MICs = 0.2-12.5 μg/ml); inhibits the growth of clinical isolates of S. aureus, E. coli, P. mirabilis, and K. pneumoniae (MICs = 0.1-25 μg/ml); protects against S. aureus, E. coli, and P. mirabilis infection in mice (ED50s = <0.09-1.78, 0.44-3.63, and 2.31-5.2 mg/animal, respectively)
Catalog No. 50-296-8921 Supplier Cayman Chemical Supplier No. 15776500 MG
A cephalosporin antibiotic; active in vitro against various Gram-positive and Gram-negative bacteria (MICs = 0.2-12.5 μg/ml); inhibits the growth of clinical isolates of S. aureus, E. coli, P. mirabilis, and K. pneumoniae (MICs = 0.1-25 μg/ml); protects against S. aureus, E. coli, and P. mirabilis infection in mice (ED50s = <0.09-1.78, 0.44-3.63, and 2.31-5.2 mg/animal, respectively)