Learn More
Medchemexpress LLC HY-W040022 25mg Medchemexpress, Cefcapene pivoxil (hydrochloride hydrate) CAS:147816-24-8 Purity:>98%
SDP
Catalog No. 502083460
Encompass_Preferred
Medchemexpress, HY-W040022 25mg Cefcapene pivoxil (hydrochloride hydrate) CAS:147816-24-8 Cefcapene pivoxil hydrochloride hydrate is a prodrug and an orally active 3rd-generation cephalosporin with broad-spectrum of anti-bacterial activity. Cefcapene pivoxil hydrochloride hydrate is used for the study of palmoplantar pustulosis (PPP) and other infectious diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3460 Supplier Medchemexpress LLC Supplier No. HYW04002225MG
Medchemexpress, HY-W040022 25mg Cefcapene pivoxil (hydrochloride hydrate) CAS:147816-24-8 Cefcapene pivoxil hydrochloride hydrate is a prodrug and an orally active 3rd-generation cephalosporin with broad-spectrum of anti-bacterial activity. Cefcapene pivoxil hydrochloride hydrate is used for the study of palmoplantar pustulosis (PPP) and other infectious diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.