Learn More
Medchemexpress LLC HY-B0136 500mg Medchemexpress, Cefdinir CAS:91832-40-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0136500MG
Encompass_Preferred
Medchemexpress, HY-B0136 500mg Cefdinir CAS:91832-40-5 Cefdinir (FK-482) is a semi-synthetic, broad-spectrum antibiotic in the third generation of the cephalosporin class, which is proved to be effective for infections caused by several Gram-negative and Gram-positive bacteria. Cefdinir can be used for the research of common bacterial infections of the ear, sinus, throat, and skin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2377