Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefdinir, G-Biosciences
SCI_ED

Catalog No. S162509
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S162509 1 g
S139715 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S162509 Supplier G-Biosciences Supplier No. RC915
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cefdinir is a third generation cephalosporin antibiotic with resistance to many Beta-lactamase enzymes. It is effective against gram-positive and gram-negative bacteria.

Cefdinir has been used to study infections caused by Staphylococcus aureus and Neisseria gonorrhoeae.

Chemical Identifiers

Molecular Weight (g/mol) 395.42

Specifications

Physical Form Powder
Quantity 1 g
Product Line Gibco
Boiling Point 708.82°C
Percent Purity 0.97
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.