Learn More
Apexbio Technology LLC Cefditoren Pivoxil 117467-28-4 100mg
SDP
Catalog No. 5000092427
Encompass_Preferred
Cefditoren pivoxil is a small-molecule prodrug antibiotic targeting penicillin-binding proteins It is designed to inhibit these proteins thereby disrupting peptidoglycan cross-linking essential for bacterial cell wall integrity Cefditoren pivoxil exerts its biological activity primarily through inhibition of bacterial cell wall synthesis This mechanism confers bactericidal activity against a broad range of Gram-positive and Gram-negative organisms Based on these pharmacological properties cefditoren pivoxil holds research potential in studying bacterial susceptibility profiles characterizing resistance mechanisms in pathogens and evaluating pharmacokinetic-pharmacodynamic relationships associated with cephalosporins
Catalog No. 50-000-92427 Supplier Apexbio Technology LLC Supplier No. B2086100
Cefditoren pivoxil is a small-molecule prodrug antibiotic targeting penicillin-binding proteins It is designed to inhibit these proteins thereby disrupting peptidoglycan cross-linking essential for bacterial cell wall integrity Cefditoren pivoxil exerts its biological activity primarily through inhibition of bacterial cell wall synthesis This mechanism confers bactericidal activity against a broad range of Gram-positive and Gram-negative organisms Based on these pharmacological properties cefditoren pivoxil holds research potential in studying bacterial susceptibility profiles characterizing resistance mechanisms in pathogens and evaluating pharmacokinetic-pharmacodynamic relationships associated with cephalosporins