Learn More
Medchemexpress LLC Cefditoren Pivoxil | 117467-28-4 | 99.29% | 620.72 | 1 ML
SDP
Catalog No. 5000386740
Encompass_Preferred
Cefditoren Pivoxil is a broad-spectrum, third-generation, oral cephalosporin antibacterial. It shows enhanced stability against many common β lactamases and has activity against both Gram-negative and Gram-positive organisms. It is used in research for infectious diseases such as acute exacerbations of chronic bronchitis, community-acquired pneumonia (CAP), streptococcal pharyngitis/tonsillitis, or uncomplicated skin and skin structure infections.
  • Enhanced stability against many common β lactamases
  • Active against Gram-negative and Gram-positive organisms
  • Used in research for various infectious diseases
Catalog No. 50-003-86740 Supplier Medchemexpress LLC Supplier No. HY17452A10MM/1ML
Cefditoren Pivoxil is a broad-spectrum, third-generation, oral cephalosporin antibacterial. It shows enhanced stability against many common β lactamases and has activity against both Gram-negative and Gram-positive organisms. It is used in research for infectious diseases such as acute exacerbations of chronic bronchitis, community-acquired pneumonia (CAP), streptococcal pharyngitis/tonsillitis, or uncomplicated skin and skin structure infections.
  • Enhanced stability against many common β lactamases
  • Active against Gram-negative and Gram-positive organisms
  • Used in research for various infectious diseases