Learn More
Medchemexpress LLC Cefditoren Pivoxil | 117467-28-4 | 99.3% | 620.72 | 5 MG
SDP
Catalog No. 5000386681
Encompass_Preferred
Cefditoren Pivoxil (ME 1207) is a broad-spectrum, third-generation, oral cephalosporin antibacterial with enhanced stability against many common β lactamases. It is active against Gram-negative and Gram-positive organisms. It can be used in the research of infectious diseases such as acute exacerbations of chronic bronchitis, community-acquired pneumonia (CAP), streptococcal pharyngitis/tonsillitis, or uncomplicated skin and skin structure infections.
  • Broad-spectrum antibacterial activity
  • Third-generation oral cephalosporin
  • Enhanced stability against many common β lactamases
  • Active against Gram-negative organisms
  • Active against Gram-positive organisms
  • Useful in research for various infection diseases
Catalog No. 50-003-86681 Supplier Medchemexpress LLC Supplier No. HY17452A5MG
Cefditoren Pivoxil (ME 1207) is a broad-spectrum, third-generation, oral cephalosporin antibacterial with enhanced stability against many common β lactamases. It is active against Gram-negative and Gram-positive organisms. It can be used in the research of infectious diseases such as acute exacerbations of chronic bronchitis, community-acquired pneumonia (CAP), streptococcal pharyngitis/tonsillitis, or uncomplicated skin and skin structure infections.
  • Broad-spectrum antibacterial activity
  • Third-generation oral cephalosporin
  • Enhanced stability against many common β lactamases
  • Active against Gram-negative organisms
  • Active against Gram-positive organisms
  • Useful in research for various infection diseases