Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefditoren Pivoxil, G-Biosciences
SCI_ED

Catalog No. S139798
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139798 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139798 Supplier G-Biosciences Supplier No. RC917
Add to Cart
Add to Cart

Cefditoren pivoxil is a Beta-lactam, cephalosporin antibiotic that has shown to be effective against an array of gram positive and negative bacteria.

In vitro studies have exhibited cefditoren pixovil to have strong activity towards methicillin and penicillin-susceptible pathogens as well as penicillin-resistant Streptococcus pneumoniae.

Chemical Identifiers

CAS 117467-28-4
Molecular Weight (g/mol) 620.72 g/mol

Specifications

Technique HPLC, Spectrometry, UV/Vis, Ion Chromatography, Mass Spectrometry
Certifications/Compliance ISO, ISO 9001, REACH, GHS/OSHA, NIST
Type Drug Standard
Element Multi-element
Industry Type Biotech
Quantity 1 g
Percent Purity >95%
Grade Laboratory
Chemical Name or Material Cefditoren pivoxil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.