Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefditoren Sodium, G-Biosciences
SCI_ED

Catalog No. S139750
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139750 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139750 Supplier G-Biosciences Supplier No. RC918
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cefditoren sodium is sodium salt of a Beta-lactam cephalosporin antibiotic that disrupts peptidoglycan synthesis of bacterial cell walls.

Cefditoren sodium has been used in comparative studies to ascertain effectiveness of newly synthesized antibiotics as well as in vitro testing of antibiotic resistance in bacteria.

Chemical Identifiers

CAS 104146-53-4
Molecular Weight (g/mol) 528.56 g/mol

Specifications

Technique HPLC, Spectrometry, UV/Vis, Ion Chromatography, Mass Spectrometry
Certifications/Compliance ISO, ISO 9001, REACH, GHS/OSHA, NIST
Type Drug Standard
Element Multi-element
Industry Type Biotech
Quantity 1 g
Percent Purity 98%
Grade Antibiotic
Chemical Name or Material Cefditoren sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.