Learn More
Cayman Chemical Cefditoren Pivoxil, Purity: ≥95%, CAS: 117467-28-4, 100 mg
SDP
Catalog No. 502990629
Encompass_Preferred
A prodrug form of cefditoren; effective against systemic S. aureus, E. coli, K. pneumoniae, P. mirabilis, or S. marcescens infections in mice (ED50s = 10, 2.5, 11, 4.4, and 11 mg/kg, respectively)
Catalog No. 50-299-0629 Supplier Cayman Chemical Supplier No. 30315100 MG
A prodrug form of cefditoren; effective against systemic S. aureus, E. coli, K. pneumoniae, P. mirabilis, or S. marcescens infections in mice (ED50s = 10, 2.5, 11, 4.4, and 11 mg/kg, respectively)