Learn More
Selleck Chemical LLC Cefepime Dihydrochloride Monohydrate S3655-25mg
SDP
Catalog No. 503110540
Encompass_Preferred
Cefepime is a cephalosporin antibacterial drug used to treat pneumonia urinary tract skin and intra-abdominal infections
Catalog No. 50-311-0540 Supplier Selleck Chemical LLC Supplier No. S365525MG
Cefepime is a cephalosporin antibacterial drug used to treat pneumonia urinary tract skin and intra-abdominal infections