Learn More
Medchemexpress LLC Cefepime | 88040-23-7 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0692100MG
Encompass_Preferred
Cefepime is a broad-spectrum cephalosporin that crosses the blood-brain barrier. It demonstrates antibacterial effects against both Gram-positive and Gram-negative aerobic bacteria and is known to induce neurotoxicity. It exerts its antibacterial effects by binding to penicillin-binding proteins.
  • Broad-spectrum cephalosporin
  • Crosses the blood-brain barrier
  • Exhibits antibacterial effects against Gram-positive and Gram-negative aerobic bacteria
  • Binds to penicillin-binding proteins
  • Known to induce neurotoxicity
Catalog No. 50-004-07631