Learn More
Medchemexpress LLC Cefepime | 88040-23-7 | 99.9% | 50 MG
SDP
Catalog No. 5000409355
Encompass_Preferred
Cefepime is a broad-spectrum cephalosporin that effectively crosses the blood-brain barrier. It demonstrates antibacterial activity against both Gram-positive and Gram-negative aerobic bacteria, functioning by binding to penicillin-binding proteins. This compound is also known to induce neurotoxicity.
  • Broad-spectrum cephalosporin
  • Crosses the blood-brain barrier
  • Antibacterial against gram-positive and gram-negative aerobic bacteria
  • Binds to penicillin-binding proteins
  • Induces neurotoxicity
Catalog No. 50-004-09355 Supplier Medchemexpress LLC Supplier No. HYB069250MG
Cefepime is a broad-spectrum cephalosporin that effectively crosses the blood-brain barrier. It demonstrates antibacterial activity against both Gram-positive and Gram-negative aerobic bacteria, functioning by binding to penicillin-binding proteins. This compound is also known to induce neurotoxicity.
  • Broad-spectrum cephalosporin
  • Crosses the blood-brain barrier
  • Antibacterial against gram-positive and gram-negative aerobic bacteria
  • Binds to penicillin-binding proteins
  • Induces neurotoxicity