Learn More
Medchemexpress LLC Cefepime | 88040-23-7 | MFCD00864890 | 99.9% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0692500MG
Encompass_Preferred
Cefepime is a broad-spectrum cephalosporin capable of crossing the blood-brain barrier, demonstrating antibacterial effects against both Gram-positive and Gram-negative aerobic bacteria. It primarily acts by binding to penicillin-binding proteins. Caution is advised as cefepime can induce neurotoxicity.
  • Broad-spectrum cephalosporin
  • Crosses the blood-brain barrier
  • Antibacterial against Gram-positive and Gram-negative aerobic bacteria
  • Exerts effects by binding to penicillin-binding proteins
  • Soluble in DMSO and H2O
Catalog No. 50-004-09354