Learn More
Medchemexpress LLC Cefepime-d3 (sulfate) | 107648-78-2 | 99.5% | 581.66 g/mol | C19H23D3N6O9S3 | 1 MG
SDP
Catalog No. 5000283918
Encompass_Preferred
Cefepime-d3 (sulfate) is a deuterium-labeled analogue of the antibiotic cefepime provided as a small, pre-weighed sulfate salt for analytical and research applications. It is intended for use as an internal standard or tracer in mass spectrometry workflows and method development; not for human or veterinary use.
  • Deuterium-labeled internal standard suitable for LC-MS/MS quantitation.
  • High purity (99.5%) for reliable analytical performance.
  • Supplied in small pre-weighed vials (e.g., 1 mg) for method development.
  • Sulfate salt form that matches analyte chemistry.
  • Suitable for pharmacokinetic, bioanalysis, and tracer studies.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-83918 Supplier Medchemexpress LLC Supplier No. HYW6541001MG
Cefepime-d3 (sulfate) is a deuterium-labeled analogue of the antibiotic cefepime provided as a small, pre-weighed sulfate salt for analytical and research applications. It is intended for use as an internal standard or tracer in mass spectrometry workflows and method development; not for human or veterinary use.
  • Deuterium-labeled internal standard suitable for LC-MS/MS quantitation.
  • High purity (99.5%) for reliable analytical performance.
  • Supplied in small pre-weighed vials (e.g., 1 mg) for method development.
  • Sulfate salt form that matches analyte chemistry.
  • Suitable for pharmacokinetic, bioanalysis, and tracer studies.
  • Intended for research use only; not for human or veterinary use.