Learn More
Apexbio Technology LLC Cefepime Dihydrochloride Monohydrate(Synonyms: Cefepime hydrochloride, Cefepime HCl, Maxipime, Cefepime hydrochloride hydrate), 500mg, CAS: 123171-59-5.
SDP
Catalog No. 502240426
Encompass_Preferred
Cefepime Dihydrochloride Monohydrate (CAS 123171-59-5) is a cephalosporin antibiotic that acts by inhibiting bacterial cell wall synthesis disrupting the structural integrity and causing cell death Experimental data indicate antimicrobial activity against both Gram-positive and Gram-negative bacteria This compound is utilized in clinical investigations targeting complex urinary tract infections severe pneumonia and multidrug-resistant bacterial infections
This item is not returnable. View return policy
Catalog No. 50-224-0426 Supplier Apexbio Technology LLC Supplier No. B3519500
Cefepime Dihydrochloride Monohydrate (CAS 123171-59-5) is a cephalosporin antibiotic that acts by inhibiting bacterial cell wall synthesis disrupting the structural integrity and causing cell death Experimental data indicate antimicrobial activity against both Gram-positive and Gram-negative bacteria This compound is utilized in clinical investigations targeting complex urinary tract infections severe pneumonia and multidrug-resistant bacterial infections