Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefepime, G-Biosciences
SCI_ED

Catalog No. S880125
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S880125 50 mg
S000038 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S880125 Supplier G-Biosciences Supplier No. RC920
Add to Cart
Add to Cart

Cefepime is a fourth generation cephalosporin antibiotic effective against gram-negative and gram-positive bacteria.

It is resistant to a variety of Beta-lactamases, and is effective against some naturally resistant bacterial species such as Pseudomonas aeruginosa, Staphylococcus aureus, and Streptococcus pneumoniae.

Chemical Identifiers

CAS 88040-23-7
Molecular Weight (g/mol) 480.56 g/mol

Specifications

Technique HPLC, Spectrometry, Calibration, Gas Chromatography, UV/Vis
Certifications/Compliance ISO, ISP 9001, REACH, GHS/OSHA, ASTM 665
Type Drug Standard
Element Multi-element
Industry Type Biotech
Quantity 50 mg
Percent Purity >95%
Grade Molecular Biology
Chemical Name or Material cefepime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.