Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefepime HCl, G-Biosciences
SCI_ED

Catalog No. S162512
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S162512 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S162512 Supplier G-Biosciences Supplier No. RC921
Add to Cart
Add to Cart

Cefepime is a fourth generation cephalosporin antibiotic effective against gram-negative and gram-positive bacteria.

It is effective against some naturally resistant bacterial species such as Pseudomonas aeruginosa, Staphylococcus aureus, and Streptococcus pneumoniae.

Chemical Identifiers

CAS 123171-59-5
Molecular Weight (g/mol) 571.50 g/mol

Specifications

Technique HPLC, Spectrometry, UV/Vis, Ion Chromatography, Mass Spectrometry
Certifications/Compliance ISO, ISO 9001, REACH, GHS/OSHA, NIST
Type Drug Standard
Element Multi-element
Industry Type Biotech
Quantity 100 mg
Percent Purity 95%
Grade Molecular Biology
Chemical Name or Material Cefepime HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.