Learn More
U.S. Pharmacopeia Cefepime Hydrochloride, 123171-59-5, MFCD06795759, 500mg
Catalog No. NC9229821
Encompass
Molecular formula C19H25ClN6O5S2 . HCl . H2O, Molecular weight 571.5, Melting Point 302 °F (150 °C) (decomposes), Synonyms: Cefepime dihydrochloride monohydrate
Catalog No. NC9229821 Supplier U.S. Pharmacopeia Supplier No. 1097636
Molecular formula C19H25ClN6O5S2 . HCl . H2O, Molecular weight 571.5, Melting Point 302 °F (150 °C) (decomposes), Synonyms: Cefepime dihydrochloride monohydrate