Learn More
Medchemexpress LLC Cefetamet | 65052-63-3 | 99.1% | C14H15N5O5S2 | 5 MG
SDP
Catalog No. 5000433420
Encompass_Preferred
Cefetamet is a cephalosporin antibiotic and the active metabolite of Cefetamet pivoxil. It targets bacterial penicillin-binding protein (PBP) and exhibits significant activity against a range of Gram-negative bacteria, including Enterobacteriaceae and *Haemophilus influenzae*, as well as Gram-positive bacteria like Streptococcus. This compound effectively kills and lyses *Treponema pallidum*.
  • Cephalosporin antibiotic
  • Active against Gram-negative and Gram-positive bacteria
  • Targets bacterial penicillin-binding protein (PBP)
  • Kills and lyses *Treponema pallidum*
  • Used in research for bacterial infections
Catalog No. 50-004-33420 Supplier Medchemexpress LLC Supplier No. HYA01115MG
Cefetamet is a cephalosporin antibiotic and the active metabolite of Cefetamet pivoxil. It targets bacterial penicillin-binding protein (PBP) and exhibits significant activity against a range of Gram-negative bacteria, including Enterobacteriaceae and *Haemophilus influenzae*, as well as Gram-positive bacteria like Streptococcus. This compound effectively kills and lyses *Treponema pallidum*.
  • Cephalosporin antibiotic
  • Active against Gram-negative and Gram-positive bacteria
  • Targets bacterial penicillin-binding protein (PBP)
  • Kills and lyses *Treponema pallidum*
  • Used in research for bacterial infections